5,6-Dimethoxy-1-indanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5,6-Dimethoxy-2,3-dihydro-1H-inden-1-one
M-Clarity™ quality level = MQ100
147826-25G 147826-5G
SIAL147826-25GEA 81.6 EUR
SIAL147826-25G SIAL147826-5G
5,6-Dimethoxy-1-indanone, Sigma-Aldrich®
5,6-Dimethoxy-1-indanone
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00003790
CAS Number: 2107-69-9
EINECS: 218-287-8