Indene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1H-Indene
M-Clarity™ quality level = MQ100
I2800-1L
I2800-250ML
SIALI2800-1LEA
217
EUR
SIALI2800-1L
SIALI2800-250ML
Indene, Sigma-Aldrich®
Indene
Formula:
C₉H₈ MW: 116.16 g/mol Boiling Pt: 183 Melting Pt: –2 °C Density: 0.998 g/cm³ (20 °C) Flash Pt: 58 Storage Temperature: Ambient |
MDL Number:
MFCD00003777 CAS Number: 95-13-6 EINECS: 202-393-6 UN: 3295 ADR: 3 III Merck Index: 13,04964 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...