4-Bromoindole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Bromo-1H-indole

M-Clarity™ quality level = MQ200

524336-1ML 524336-5ML
SIAL524336-1MLEA 113 EUR
SIAL524336-1ML SIAL524336-5ML
4-Bromoindole, Sigma-Aldrich®
4-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD00671502
CAS Number: 52488-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR