5-Bromoindole
Supplier: Thermo Scientific
Synonyms:
5-Bromo-1H-indole
RH00766.10
RH00766.50
MAYBRH00766.10EA
0
EUR
MAYBRH00766.10
MAYBRH00766.50
5-Bromoindole
5-Bromoindole
Formula:
C₈H₆BrN MW: 196.05 g/mol Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005670 CAS Number: 10075-50-0 EINECS: 233-208-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...