6-Bromoindole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-Bromo-1H-indole

M-Clarity™ quality level = MQ200

524344-1G 524344-5G
SIAL524344-1GEA 85.1 EUR
SIAL524344-1G SIAL524344-5G
6-Bromoindole, Sigma-Aldrich®
6-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR