Indole-5-carbonitrile 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole
110790010 110790050
ACRO110790010EA 19.8 EUR
ACRO110790010 ACRO110790050
Indole-5-carbonitrile 99%
Indole-5-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 106…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005669
CAS Number: 15861-24-2
EINECS: 239-986-4
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 105 to 108 °C
GC ≥98.5 %
Titrable acid ≤0.001 meq/g