Indole-3-carboxylic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1H-Indole-3-carboxylic acid
M-Clarity™ quality level = MQ100
284734-1G
284734-5G
SIAL284734-1GEA
26.1
EUR
SIAL284734-1G
SIAL284734-5G
Indole-3-carboxylic acid, Sigma-Aldrich®
Indole-3-carboxylic acid
Formula:
C₉H₇NO₂ MW: 161.16 g/mol |
MDL Number:
MFCD00005624 CAS Number: 771-50-6 EINECS: 212-231-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...