6-Fluoroindole, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 6-Fluoro-1H-indole

M-Clarity™ quality level = MQ100

349593-1G 349593-5G
SIAL349593-1GEA 28.1 EUR
SIAL349593-1G SIAL349593-5G
6-Fluoroindole, Sigma-Aldrich®
6-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR