6-Aminoindole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
630721-1G
630721-5G
SIAL630721-1GEA
76.1
EUR
SIAL630721-1G
SIAL630721-5G
6-Aminoindole, Sigma-Aldrich®
6-Aminoindole
Formula:
C₈H₈N₂ MW: 132.16 g/mol Boiling Pt: 161…166 °C (2.3 mmHg) Melting Pt: 65…66 °C Storage Temperature: Freezer |
MDL Number:
MFCD02093959 CAS Number: 5318-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...