4-Nitroindole 98%

Supplier: Thermo Scientific

ACRO124380025EA 301 EUR
ACRO124380025
4-Nitroindole 98%
4-Nitroindole

Order Now

Specification Test Results

Appearance Yellow Granular powder
Infrared spectrum Conforms
Melting point 203 to 207 °C
TLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR