5-Methoxyindole, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 5-Methoxy-1H-indole, Indol-5-yl methyl ether
M-Clarity™ quality level = MQ100
M14900-1G M14900-5G
SIALM14900-1GEA 25.6 EUR
SIALM14900-1G SIALM14900-5G
5-Methoxyindole, Sigma-Aldrich®
5-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 55…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005674
CAS Number: 1006-94-6
EINECS: 213-745-3