Indole, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
I3408-100G I3408-25G I3408-500G
SIALI3408-100GEA 90 EUR
SIALI3408-100G SIALI3408-25G SIALI3408-500G
Indole, Sigma-Aldrich®
Indole
Formula: C₈H₇N
Boiling Pt: 254…255 °C (1013 hPa)
Melting Pt: 50…51 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,04985