Indole, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W259306-100G-K W259306-10KG-K W259306-1KG-K W259306-25KG-K
SIALW259306-100G-KEA 75.8 EUR
SIALW259306-100G-K SIALW259306-10KG-K SIALW259306-1KG-K SIALW259306-25KG-K
Indole, Sigma-Aldrich®
Indole
Formula: C₈H₇N
Boiling Pt: 254…255 °C (1013 hPa)
Melting Pt: 50…51 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,04985