2-Phenylindole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Phenyl-1H-indole
M-Clarity™ quality level = MQ100
P26600-100G P26600-5G
SIALP26600-100GEA 52 EUR
SIALP26600-100G SIALP26600-5G
2-Phenylindole, Sigma-Aldrich®
2-Phenylindole
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Boiling Pt: 420 °C (1013 hPa)
Melting Pt: 186…187 °C
MDL Number: MFCD00005608
CAS Number: 948-65-2
EINECS: 213-436-3