4-Bromoisoquinoline, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
B70202-10G
B70202-25G
SIALB70202-10GEA
58.8
EUR
SIALB70202-10G
SIALB70202-25G
4-Bromoisoquinoline, Sigma-Aldrich®
4-Bromoisoquinoline
Formula:
C₉H₆BrN MW: 208.06 g/mol Boiling Pt: 280 °C (1013 hPa) Melting Pt: 42…42.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006904 CAS Number: 1532-97-4 EINECS: 216-244-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...