Isoquinoline-1-amine, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Isoquinolin-1-amine, 1-Aminoisoquinoline

M-Clarity™ quality level = MQ100

178594-1G 178594-5G
SIAL178594-1GEA 34.4 EUR
SIAL178594-1G SIAL178594-5G
Isoquinoline-1-amine, Sigma-Aldrich®
Isoquinoline-1-amine
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 122…124 °C
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00024137
CAS Number: 1532-84-9
EINECS: 216-243-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR