Lanthanum(III) bromide, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

449822-10G 449822-2G
SIAL449822-10GEA 440 EUR
SIAL449822-10G SIAL449822-2G
Lanthanum(III) bromide, Sigma-Aldrich®
Lanthanum (III) bromide
Formula: Br₃La
MW: 378.62 g/mol
Melting Pt: 789 °C
Density: 5.06 g/cm³ (25 °C)
MDL Number: MFCD00016161
CAS Number: 13536-79-3
EINECS: 236-896-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR