Erbium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
Warning
ampouled under argon
35739.06 35739.14
35739.06EA 214 EUR
35739.06 35739.14
Erbium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Erbium trichloride
Purity: 99.9% CAS No.: 10138-41-7 Molecular Formula: ErCl3 Formula Weight: 273.62 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic Boiling Point: 1500° MDL No.: MFCD00010989
Formula: Cl₃Er
MW: 273.62 g/mol
Boiling Pt: 1500 °C
Melting Pt: 774 °C
Density: 4.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010989
CAS Number: 10138-41-7
EINECS: 233-385-0