Erbium trichloride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
449792-25G 449792-5G
SIAL449792-25GEA 860 EUR
SIAL449792-25G SIAL449792-5G
Erbium trichloride, Sigma-Aldrich®
Erbium trichloride
Formula: Cl₃Er
MW: 273.62 g/mol
Boiling Pt: 1500 °C (1013 hPa)
Melting Pt: 751 °C
Density: 4.1 g/cm³ (20 °C)
MDL Number: MFCD00010989
CAS Number: 10138-41-7
EINECS: 233-385-0