Europium(III) chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

429732-1G 429732-5G
SIAL429732-1GEA 114 EUR
SIAL429732-1G SIAL429732-5G
Europium(III) chloride, Sigma-Aldrich®
Europium (III) chloride
Formula: Cl₃Eu
Melting Pt: 626…638 °C
Density: 4.89 g/cm³ (20 °C)
MDL Number: MFCD00010994
CAS Number: 10025-76-0
EINECS: 233-040-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR