Holmium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Holmium trichloride
M-Clarity™ quality level = MQ100
450901-1G
450901-5G
SIAL450901-1GEA
110
EUR
SIAL450901-1G
SIAL450901-5G
Holmium(III) chloride, Sigma-Aldrich®
Holmium(III) chloride
Formula:
Cl₃Ho MW: 271.29 g/mol Boiling Pt: 1500 °C (1013 hPa) Melting Pt: 699…709 °C Density: 3.7 g/cm³ (20 °C) |
MDL Number:
MFCD00011051 CAS Number: 10138-62-2 EINECS: 233-387-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...