Lanthanum(III) chloride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Lanthanum chloride, Lanthanum trichloride
M-Clarity™ quality level = MQ100
298182-10G
298182-50G
SIAL298182-10GEA
120
EUR
SIAL298182-10G
SIAL298182-50G
Lanthanum(III) chloride, Sigma-Aldrich®
Lanthanum (III) chloride
Formula:
Cl₃La Melting Pt: 857 °C Density: 3.8773 g/cm³ (–0.15 °C) |
MDL Number:
MFCD00011068 CAS Number: 10099-58-8 EINECS: 233-237-5 Merck Index: 13,05379 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...