Neodymium trichloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Neodymium (III) chloride
M-Clarity™ quality level = MQ100
449946-25G 449946-5G
SIAL449946-25GEA 1140 EUR
SIAL449946-25G SIAL449946-5G
Neodymium trichloride, Sigma-Aldrich®
Neodymium trichloride
Formula: Cl₃Nd
MW: 250.6 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 758 °C
Density: 4.13 g/cm³ (20 °C)
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5