Terbium(III) chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

439657-10G 439657-2G
SIAL439657-10GEA 482 EUR
SIAL439657-10G SIAL439657-2G
Terbium(III) chloride, Sigma-Aldrich®
Terbium (III) chloride
Formula: Cl₃Tb
Melting Pt: 588 °C
Density: 4.35 g/cm³ (25 °C)
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 13,09087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR