Europium(II) iodide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Europium diiodide

M-Clarity™ quality level = MQ100

474770-1G 474770-5G
SIAL474770-1GEA 201 EUR
SIAL474770-1G SIAL474770-5G
Europium(II) iodide, Sigma-Aldrich®
Europium (II) iodide
Formula: EuI₂
MW: 405.77 g/mol
CAS Number: 22015-35-6
EINECS: 244-721-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR