Cerium(IV) oxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ceric oxide, Cerium dioxide

M-Clarity™ quality level = MQ100

544841-5G 544841-25G
SIAL544841-5GEA 49.6 EUR
SIAL544841-5G SIAL544841-25G
Cerium(IV) oxide, Sigma-Aldrich®
Cerium (IV) oxide
Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 7.13 g/cm³ (25 °C)
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR