Dysprosium(III) oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
203181-25G
203181-5G
SIAL203181-25GEA
366
EUR
SIAL203181-25G
SIAL203181-5G
Dysprosium(III) oxide, Sigma-Aldrich®
Dysprosium (III) oxide
Formula:
Dy₂O₃ MW: 373 g/mol Melting Pt: ∼2400 °C Density: 7.81 g/cm³ (20 °C) |
MDL Number:
MFCD00010986 CAS Number: 1308-87-8 EINECS: 215-164-0 Merck Index: 14,03482 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...