Dysprosium(III) oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

289264-100G 289264-25G
SIAL289264-100GEA 584 EUR
SIAL289264-100G SIAL289264-25G
Dysprosium(III) oxide, Sigma-Aldrich®
Dysprosium (III) oxide
Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: ∼2400 °C
Density: 7.81 g/cm³ (20 °C)
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR