Europium(III) oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
289221-25G 289221-5G
SIAL289221-25GEA 827 EUR
SIAL289221-25G SIAL289221-5G
Europium(III) oxide, Sigma-Aldrich®
Europium (III) oxide
Formula: Eu₂O₃
MW: 351.92 g/mol
Melting Pt: 2325 °C
Density: 7.42 g/cm³ (20 °C)
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904