Europium(III) oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
323543-1G
323543-5G
SIAL323543-1GEA
168
EUR
SIAL323543-1G
SIAL323543-5G
Europium(III) oxide, Sigma-Aldrich®
Europium (III) oxide
Formula:
Eu₂O₃ MW: 351.92 g/mol Melting Pt: 2325 °C Density: 7.42 g/cm³ (20 °C) |
MDL Number:
MFCD00010997 CAS Number: 1308-96-9 EINECS: 215-165-6 Merck Index: 14,03904 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...