Digadolinium trioxide 99.9% (trace metals basis)
Supplier: Thermo Scientific
Warning
Synonyms: Gadolinium (III) oxide
315510250 315510100 315511000
ACRO315510250EA 70.9 EUR
ACRO315510250 ACRO315510100 ACRO315511000
Digadolinium trioxide 99.9% (trace metals basis)
Digadolinium trioxide
Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 1900 °C
Density: 7,407 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Order Now
Specification Test Results
Appearance White Powder
Total conc. of imp.as REO ≤0.1 %