Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Gadolinium (III) oxide
11290.09 11290.18 11290.30 11290.A1
11290.09EA 47.8 EUR
11290.09 11290.18 11290.30 11290.A1
Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Digadolinium trioxide
Purity: 99.99% CAS No.: 12064-62-9 Molecular Formula: Gd2O3 Formula Weight: 362.50 Physical Form: 10 Micron Powder Density: 7.407 Melting Point: 2330° Merck Reference: 14,4326 MDL No.: MFCD00011026
Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2330 °C
Density: 7.407
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Merck Index: 14,4326