Digadolinium trioxide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Gadolinium (III) oxide
M-Clarity™ quality level = MQ100
278513-100G
278513-25G
SIAL278513-100GEA
294
EUR
SIAL278513-100G
SIAL278513-25G
Digadolinium trioxide, Sigma-Aldrich®
Digadolinium trioxide
Formula:
Gd₂O₃ MW: 362.5 g/mol Melting Pt: 2410 Density: 7.407 g/cm³ (20 °C) |
MDL Number:
MFCD00011026 CAS Number: 12064-62-9 EINECS: 235-060-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...