Digadolinium trioxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Gadolinium (III) oxide

M-Clarity™ quality level = MQ100

637335-10G 637335-50G
SIAL637335-10GEA 292 EUR
SIAL637335-10G SIAL637335-50G
Digadolinium trioxide, Sigma-Aldrich®
Digadolinium trioxide
Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2410 °C
Density: 7.407 g/cm³ (20 °C)
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR