Holmium(III) oxide ≥99.9%, GPR RECTAPUR®
Supplier: VWR Chemicals
24591.105EA 148 EUR
24591.105
Holmium(III) oxide ≥99.9%, GPR RECTAPUR®
Holmium(III) oxide
Formula: Ho₂O₃
MW: 377,86 g/mol
Boiling Pt: 2600 °C (1013 hPa)
Melting Pt: 2400 °C
Density: 3,79 g/cm³ (20 °C)
MDL Number: MFCD00011053
CAS Number: 12055-62-8
Order Now
Specification Test Results
Assay Min. 99.9 %