Holmium(III) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Diholmium trioxide, Holmium oxide
M-Clarity™ quality level = MQ100
229679-10G 229679-1G
SIAL229679-10GEA 663 EUR
SIAL229679-10G SIAL229679-1G
Holmium(III) oxide, Sigma-Aldrich®
Holmium (III) oxide
Formula: Ho₂O₃
MW: 377.86 g/mol
Boiling Pt: 2600
Melting Pt: 2400
Density: 3.79 g/cm³ (20 °C)
MDL Number: MFCD00011053
CAS Number: 12055-62-8
EINECS: 235-015-3