Holmium(III) oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL637327-10GEA 224 EUR
SIAL637327-10G
Holmium(III) oxide, Sigma-Aldrich®
Holmium(III) oxide
Formula: Ho₂O₃
MW: 377,86 g/mol
Boiling Pt: 2600 °C (1013 hPa)
Melting Pt: 2400 °C
Density: 3,79 g/cm³ (20 °C)
MDL Number: MFCD00011053
CAS Number: 12055-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR