Lanthanum(III) oxide, Sigma-Aldrich®
Supplier: Merck
L4000-1KG L4000-100G L4000-250G L4000-500G
SIALL4000-1KGEA 656 EUR
SIALL4000-1KG SIALL4000-100G SIALL4000-250G SIALL4000-500G
Lanthanum(III) oxide, Sigma-Aldrich®
Lanthanum (III) oxide
Formula: La₂O₃
MW: 325.81 g/mol
Boiling Pt: 4200 °C (1013 hPa)
Melting Pt: 2315 °C
Density: 6.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5