Samarium(III) oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

228672-100G 228672-10G
SIAL228672-100GEA 484 EUR
SIAL228672-100G SIAL228672-10G
Samarium(III) oxide, Sigma-Aldrich®
Samarium (III) oxide
Formula: O₃Sm₂
MW: 348.72 g/mol
Melting Pt: 2325
Density: 7.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS Number: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,08347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR