Safety shoes, lace-up, Basic, Jazek, Jori
Supplier: ELTEN
12520/46 12520/47 12520/48 12520/36 12520/37 12520/38 12520/39 12520/40 12520/41 12520/42 12520/43 12520/44 12520/45
ELTN12520/46EA 59.9 EUR
ELTN12520/46 ELTN12520/47 ELTN12520/48 ELTN12520/36 ELTN12520/37 ELTN12520/38 ELTN12520/39 ELTN12520/40 ELTN12520/41 ELTN12520/42 ELTN12520/43 ELTN12520/44 ELTN12520/45
Safety shoes, lace-up, Basic, Jazek, Jori
Shoes and Boots
Durable safety shoes made from cowhide leather and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), reflective materials, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU New Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/blue
Certifikace: EN ISO 20345; S2, SRC.