Naphthacene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,3-Benzanthracene, Benz[b]anthracene, Tetracene
M-Clarity™ quality level = MQ100
B2403-100MG
B2403-1G
B2403-500MG
SIALB2403-100MGEA
66.6
EUR
SIALB2403-100MG
SIALB2403-1G
SIALB2403-500MG
Naphthacene, Sigma-Aldrich®
Naphthacene
Formula:
C₁₈H₁₂ MW: 228.29 g/mol Boiling Pt: 437 °C (1013 hPa) Melting Pt: 356 °C Density: 1.323 g/cm³ (20 °C) |
MDL Number:
MFCD00003702 CAS Number: 92-24-0 EINECS: 202-138-9 Merck Index: 12,06456 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...