2,6-Naphthalenedicarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,6-Naphthalenedicarboxylic Acid, Naphthalene-2,6-dicarboxylic acid
MQ100
301353-25G 301353-5G
SIAL301353-25GEA 111 EUR
SIAL301353-25G SIAL301353-5G
2,6-Naphthalenedicarboxylic acid, Sigma-Aldrich®
2,6-Naphthalenedicarboxylic acid
Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Boiling Pt: 422
Melting Pt: 366…368 °C
MDL Number: MFCD00004105
CAS Number: 1141-38-4
EINECS: 214-527-0