1-Iodonaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
238139-10G 238139-50G
SIAL238139-10GEA 87.5 EUR
SIAL238139-10G SIAL238139-50G
1-Iodonaphthalene, Sigma-Aldrich®
1-Iodonaphthalene
Formula: C₁₀H₇I
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.738 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003876
CAS Number: 90-14-2
EINECS: 201-968-9