1,5-Diaminonaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,5-naphthylenediamine, 1,5-Naphthalenediamine
MQ100
D21200-100G D21200-25G
SIALD21200-100GEA 143 EUR
SIALD21200-100G SIALD21200-25G
1,5-Diaminonaphthalene, Sigma-Aldrich®
1,5-Diaminonaphthalene
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 381
Melting Pt: 188
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004029
CAS Number: 2243-62-1
EINECS: 218-817-8
UN: 3077
ADR: 9 III