1,6-Dihydroxynaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Naphthalene-1,6-diol
M-Clarity™ quality level = MQ200
274127-250G 274127-50G
SIAL274127-250GEA 385 EUR
SIAL274127-250G SIAL274127-50G
1,6-Dihydroxynaphthalene, Sigma-Aldrich®
1,6-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 135.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003981
CAS Number: 575-44-0
EINECS: 209-386-7