2,7-Dihydroxynaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol
M-Clarity™ quality level = MQ200
D116408-100G D116408-5G
SIALD116408-100GEA 114 EUR
SIALD116408-100G SIALD116408-5G
2,7-Dihydroxynaphthalene, Sigma-Aldrich®
2,7-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS Number: 582-17-2
EINECS: 209-478-7