2-Vinylnaphthalene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

V2909-25G V2909-5G
SIALV2909-25GEA 457 EUR
SIALV2909-25G SIALV2909-5G
2-Vinylnaphthalene, Sigma-Aldrich®
2-Vinylnaphthalene
Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 135…137 °C (18 mmHg)
Melting Pt: 66
Storage Temperature: Refrigerator
MDL Number: MFCD00004125
CAS Number: 827-54-3
EINECS: 212-573-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR