1-Methylnaphthalene, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: α-Methylnaphthalene

M-Clarity™ quality level = MQ100

M56808-100G M56808-500G M56808-5G
SIALM56808-100GEA 41.6 EUR
SIALM56808-100G SIALM56808-500G SIALM56808-5G
1-Methylnaphthalene, Sigma-Aldrich®
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 244…245 °C (1013 hPa)
Melting Pt: –33…–32 °C
Density: 1.022 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR