1-Methylnaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: α-Methylnaphthalene
M-Clarity™ quality level = MQ300
W319309-100G-K W319309-1KG-K W319309-5KG-K
SIALW319309-100G-KEA 114 EUR
SIALW319309-100G-K SIALW319309-1KG-K SIALW319309-5KG-K
1-Methylnaphthalene, Sigma-Aldrich®
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 244…245 °C (1013 hPa)
Melting Pt: –33…–32 °C
Density: 1.022 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III