Copper(II) bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cupric bromide, Cuprous bromide
M-Clarity™ quality level = MQ100
437867-25G 437867-5G
SIAL437867-25GEA 255 EUR
SIAL437867-25G SIAL437867-5G
Copper(II) bromide, Sigma-Aldrich®
Copper (II) bromide
Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900
Melting Pt: 498
Density: 4.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8 III
Merck Index: 13,02656